Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003712.1 | NP_001003712.1 | 15 | 2108 | ||
NM_001319652.1 | NP_001306581.1 | 18 | 2233 | |||
NM_001319653.1 | NP_001306582.1 | 16 | 2145 | |||
NM_001319655.1 | NP_001306584.1 | 13 | 2039 | |||
NM_020841.4 | NP_065892.1 | 16 | 2154 | |||
XM_005268621.4 | XP_005268678.1 | 19 | 2252 | |||
XM_006719224.2 | XP_006719287.1 | 15 | 2113 | |||
XM_011537855.2 | XP_011536157.1 | 16 | 1771 | |||
XM_011537856.2 | XP_011536158.1 | 17 | 2189 | |||
XM_011537857.2 | XP_011536159.1 | 19 | 2243 | |||
XM_017018768.1 | XP_016874257.1 | 16 | 1786 | |||
XM_017018769.1 | XP_016874258.1 | 17 | 2157 | |||
GenBank mRNA | AB040884.3 | - | 16 | 1825 | ||
AF323730.1 | - | 15 | 2108 | |||
AF392452.1 | - | 16 | 2155 | |||
AK092160.1 | - | 13 | 2040 | |||
AK289997.1 | - | 16 | 1874 | |||
AL832876.1 | - | 3 | 210 | |||
BC012935.1 | - | 2 | 139 | |||
BC082235.1 | - | 16 | 2093 | |||
BC093834.1 | - | 15 | 1753 | 7939679 | ||
BC101529.1 | - | 15 | 1753 | 8069035 | ||
BC111728.1 | - | 16 | 2048 | 40029118 | ||
BX647211.1 | - | 15 | 1710 | |||
CR749542.1 | - | 18 | 1958 |