Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144063.1 | NP_001137535.1 | 8 | 853 | ||
NM_020896.3 | NP_065947.1 | 9 | 1057 | |||
NM_145638.2 | NP_663613.1 | 8 | 846 | |||
XM_011519873.2 | XP_011518175.1 | 9 | 1396 | |||
XM_017017163.1 | XP_016872652.1 | 9 | 1404 | |||
XM_017017164.1 | XP_016872653.1 | 4 | 975 | |||
GenBank mRNA | AB074006.1 | - | 9 | 998 | ||
AF392453.1 | - | 9 | 970 | |||
AF410430.1 | - | 8 | 915 | |||
AK055617.1 | - | 4 | 391 | |||
AK056510.1 | - | 9 | 1015 | |||
AK293000.1 | - | 8 | 802 | |||
AK301003.1 | - | 7 | 693 | |||
AK303205.1 | - | 9 | 875 | |||
AK309411.1 | - | 9 | 1035 | |||
AL136918.1 | - | 9 | 970 | |||
BC032646.1 | - | 8 | 767 | 5518491 | ||
BC039579.1 | - | 9 | 968 | 5769002 |