Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144063.1 | NP_001137535.1 | 12 | 1435 | ||
NM_020896.3 | NP_065947.1 | 13 | 1639 | |||
NM_145638.2 | NP_663613.1 | 12 | 1428 | |||
XM_011519873.2 | XP_011518175.1 | 13 | 1978 | |||
XM_017017163.1 | XP_016872652.1 | 13 | 1986 | |||
XM_017017164.1 | XP_016872653.1 | 8 | 1557 | |||
GenBank mRNA | AB074006.1 | - | 13 | 1580 | ||
AF392453.1 | - | 13 | 1552 | |||
AF410430.1 | - | 12 | 1497 | |||
AK055617.1 | - | 8 | 973 | |||
AK056510.1 | - | 13 | 1597 | |||
AK293000.1 | - | 12 | 1384 | |||
AK297475.1 | - | 9 | 734 | |||
AK301003.1 | - | 11 | 1275 | |||
AK303205.1 | - | 13 | 1457 | |||
AK309411.1 | - | 13 | 1617 | |||
AL136918.1 | - | 13 | 1552 | |||
BC017505.1 | - | 4 | 295 | |||
BC032646.1 | - | 12 | 1349 | 5518491 | ||
BC039579.1 | - | 13 | 1550 | 5769002 |