Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144063.1 | NP_001137535.1 | 21 | 3512 | ||
NM_020896.3 | NP_065947.1 | 22 | 3716 | |||
NM_145638.2 | NP_663613.1 | 21 | 3505 | |||
XM_011519873.2 | XP_011518175.1 | 22 | 4055 | |||
XM_017017163.1 | XP_016872652.1 | 22 | 4063 | |||
XM_017017164.1 | XP_016872653.1 | 17 | 3634 | |||
GenBank mRNA | AB074006.1 | - | 22 | 3669 | ||
AF392453.1 | - | 22 | 3640 | |||
AK055617.1 | - | 17 | 3061 | |||
AK056510.1 | - | 22 | 3675 | |||
AL136918.1 | - | 22 | 3640 | |||
BC032646.1 | - | 21 | 3438 | 5518491 | ||
BC039579.1 | - | 22 | 3639 | 5769002 |