Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012964.2 | NP_001012982.1 | 6 | 1598 | ||
NM_001012965.2 | NP_001012983.1 | 5 | 1550 | |||
NM_001319948.1 | NP_001306877.1 | 6 | 1244 | |||
NM_001319949.1 | NP_001306878.1 | 5 | 1441 | |||
NM_002774.3 | NP_002765.1 | 7 | 1401 | |||
GenBank mRNA | AF013988.1 | - | 7 | 1303 | ||
AY279383.1 | - | 6 | 1245 | |||
AY318867.1 | - | 6 | 1407 | |||
AY318869.1 | - | 6 | 1393 | |||
BC015525.2 | - | 6 | 1374 | 3852344 | ||
D78203.1 | - | 7 | 1312 |