Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322977.1 | NP_001309906.1 | 2 | 257 | ||
NM_001322978.1 | NP_001309907.1 | 2 | 229 | |||
NM_001322980.1 | NP_001309909.1 | 2 | 229 | |||
NM_001322981.1 | NP_001309910.1 | 2 | 257 | |||
NM_001322982.1 | NP_001309911.1 | 2 | 257 | |||
NM_001322983.1 | NP_001309912.1 | 2 | 229 | |||
NM_022754.6 | NP_073591.2 | 2 | 229 | |||
GenBank mRNA | AF327346.1 | - | 2 | 139 | ||
AK022287.1 | - | 2 | 320 | |||
AK056915.1 | - | 2 | 134 | |||
BC020517.1 | - | 1 | 24 | 3888861 | ||
BC063241.1 | - | 2 | 147 | 6187994 | ||
BP199230.1 | - | 2 | 257 | |||
BX648188.1 | - | 2 | 145 | |||
DA753705.1 | - | 2 | 216 |