Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020002.1 | XP_016875491.1 | 2 | 662 | ||
GenBank mRNA | AF212239.1 | - | 2 | 274 | ||
AK096947.1 | - | 1 | 573 | |||
AK223378.1 | - | 2 | 289 | |||
AK311977.1 | - | 2 | 287 | |||
AY007436.1 | - | 2 | 270 | |||
BC029355.1 | - | 2 | 291 | 4613026 |