Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178034.3 | NP_828848.3 | 20 | 3514 | ||
XM_011521467.2 | XP_011519769.1 | 19 | 3345 | |||
GenBank mRNA | AB090876.1 | - | 20 | 3517 | ||
BC034571.1 | - | 5 | 2572 |