Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006327.3 | NP_006318.1 | 6, 7 | 686 | ||
NR_073030.1 | 5, 6 | 627 | ||||
GenBank mRNA | AF030162.1 | - | 6, 7 | 638 | ||
AK223331.1 | - | 6, 7 | 626 | |||
AK304883.1 | - | 5, 6 | 510 | |||
AY442341.1 | - | 6, 7 | 638 | |||
BC062707.1 | - | 6, 7 | 687 | 4772587 | ||
BC066951.1 | - | 6, 7 | 636 | 5264814 | ||
BY799488.2 | - | 5, 6 | 560 |