Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002792.3 | NP_002783.1 | 6 | 767 | ||
GenBank mRNA | AF022815.1 | - | 6 | 636 | ||
AF054185.1 | - | 6 | 697 | |||
AK127210.1 | - | 2 | 3206 | |||
AK312025.1 | - | 6 | 662 | |||
BC001895.1 | - | 6 | 700 | |||
BC004427.1 | - | 6 | 685 | 2819923 | ||
BC014999.1 | - | 6 | 695 | |||
BI258951.1 | - | 6 | 772 | |||
BT007165.1 | - | 6 | 612 | |||
GQ472224.1 | - | 6 | 685 |