Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000113.2 | NP_000104.1 | 2 | 475 | ||
GenBank mRNA | AF007871.1 | - | 2 | 440 | ||
AK075343.1 | - | 2 | 425 | |||
AK294677.1 | - | 2 | 565 | |||
AK314505.1 | - | 2 | 419 | |||
BC000674.2 | - | 2 | 436 | 3347611 | ||
BC014484.1 | - | 2 | 417 | 4869856 | ||
BT006931.1 | - | 2 | 398 |