Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002795.3 | NP_002786.2 | 6 | 698 | ||
NR_104194.1 | 5 | 520 | ||||
NR_104195.1 | 6 | 621 | ||||
GenBank mRNA | BC013008.2 | - | 6 | 661 | 3010175 | |
BG184666.1 | - | 2 | 114 | |||
BQ428285.1 | - | 6 | 581 | |||
CN412852.1 | - | 5 | 424 | |||
D26598.1 | - | 6 | 601 |