Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000465.3 | NP_000456.2 | 3 | 480 | ||
NM_001282543.1 | NP_001269472.1 | 2 | 423 | |||
NM_001282549.1 | NP_001269478.1 | 3 | 480 | |||
NR_104216.1 | 3 | 480 | ||||
XM_017004613.1 | XP_016860102.1 | 4 | 528 | |||
XM_017004614.1 | XP_016860103.1 | 4 | 528 | |||
GenBank mRNA | AK223409.1 | - | 3 | 474 | ||
AK314260.1 | - | 3 | 440 | |||
AX794378.1 | - | 3 | 411 | |||
BC126426.1 | - | 3 | 408 | 8992142 | ||
BC126428.1 | - | 3 | 408 | 8992144 | ||
DB088853.1 | - | 3 | 481 | |||
JF790280.1 | - | 2 | 354 | |||
JF790282.1 | - | 3 | 411 | |||
JF790285.1 | - | 3 | 366 | |||
JF790287.1 | - | 3 | 366 | |||
U76638.1 | - | 3 | 411 |