Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000465.3 | NP_000456.2 | 4, 5 | 1464 | ||
NM_001282543.1 | NP_001269472.1 | 3, 4 | 1407 | |||
NR_104212.1 | 3, 4 | 1315 | ||||
NR_104215.1 | 2, 3 | 1258 | ||||
XM_017004613.1 | XP_016860102.1 | 5, 6 | 1512 | |||
XM_017004614.1 | XP_016860103.1 | 5, 6 | 1512 | |||
GenBank mRNA | AK223409.1 | - | 4, 5 | 1458 | ||
AK301843.1 | - | 3, 4 | 1277 | |||
AK314260.1 | - | 4, 5 | 1424 | |||
AX794378.1 | - | 4, 5 | 1395 | |||
BC126426.1 | - | 4, 5 | 1392 | 8992142 | ||
BC126428.1 | - | 4, 5 | 1392 | 8992144 | ||
JF790280.1 | - | 3, 4 | 1338 | |||
JF790281.1 | - | 2, 3 | 1189 | |||
JF790287.1 | - | 4, 5 | 942 | |||
U76638.1 | - | 4, 5 | 1395 |