Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000575.4 | NP_000566.3 | 4 | 1181 | ||
GenBank mRNA | AK314850.1 | - | 4 | 276 | ||
BC013142.2 | - | 3 | 226 | 3875617 | ||
BT007014.1 | - | 3 | 217 | |||
CR457414.1 | - | 3 | 217 | |||
M15329.1 | - | 3 | 222 | |||
X02531.1 | - | 3 | 217 | |||
X02851.1 | - | 4 | 275 | |||
X56086.1 | - | 4 | 243 |