Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000893.3 | NP_000884.1 | 2 | 451 | ||
NM_001102416.2 | NP_001095886.1 | 2 | 451 | |||
NM_001166451.1 | NP_001159923.1 | 2 | 451 | |||
GenBank mRNA | AI133186.1 | - | 2 | 361 | ||
AK223589.1 | - | 2 | 400 | |||
AK290839.1 | - | 2 | 350 | |||
AK298440.1 | - | 3 | 365 | |||
AK303768.1 | - | 2 | 431 | |||
BC026253.1 | - | 2 | 220 | |||
BC060039.1 | - | 2 | 354 | 4593077 | ||
BG430928.1 | - | 2 | 430 | |||
CD014130.1 | - | 2 | 413 | |||
DA537547.1 | - | 2 | 429 | |||
DC362362.1 | - | 2 | 451 | |||
K02566.1 | - | 2 | 266 |