Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002802.2 | NP_002793.2 | 4, 5 | 338 | ||
XM_017021470.1 | XP_016876959.1 | 4, 5 | 1857 | |||
XM_017021471.1 | XP_016876960.1 | 5, 6 | 414 | |||
GenBank mRNA | AB451311.1 | - | 1 | 290 | ||
AB451441.1 | - | 1 | 290 | |||
AK222668.1 | - | 4, 5 | 306 | |||
AK299121.1 | - | 3, 4 | 214 | |||
AK313325.1 | - | 4, 5 | 327 | |||
BC000512.2 | - | 1 | 324 | 2822718 | ||
BC016368.1 | - | 1 | 318 | 4133348 | ||
BC018859.2 | - | 1 | 291 | |||
BC067741.1 | - | 1 | 311 | 5264945 | ||
BC073818.1 | - | 1 | 296 | 5456334 | ||
BT009826.1 | - | 1 | 290 | |||
CR457044.1 | - | 1 | 290 | |||
L02426.1 | - | 4, 5 | 350 |