Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001357.4 | NP_001348.2 | 28 | 4136 | ||
NR_033302.1 | 29 | 4273 | ||||
GenBank mRNA | AK096582.1 | - | 9 | 2366 | ||
AK130564.1 | - | 4 | 1032 | |||
AK226102.1 | - | 29 | 4254 | |||
BC058896.1 | - | 7 | 1359 | |||
BC107881.1 | - | 8 | 1480 | |||
L13848.1 | - | 28 | 4032 | |||
U03643.1 | - | 7 | 1215 |