Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253835.1 | NP_001240764.1 | 2 | 581 | ||
NM_001553.2 | NP_001544.1 | 2 | 581 | |||
GenBank mRNA | AF540057.1 | - | 3 | 310 | ||
AK303915.1 | - | 2 | 583 | |||
AK316082.1 | - | 2 | 581 | |||
BC017201.2 | - | 2 | 569 | 3632247 | ||
BC066339.1 | - | 2 | 581 | 5296697 | ||
BT006654.1 | - | 2 | 547 | |||
L19182.1 | - | 2 | 560 | |||
S75725.1 | - | 2 | 569 |