Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001789.2 | NP_001780.2 | 15 | 2552 | ||
NM_201567.1 | NP_963861.1 | 14 | 2432 | |||
XM_006713434.2 | XP_006713497.1 | 15 | 2644 | |||
XM_006713435.2 | XP_006713498.1 | 14 | 2524 | |||
XM_011534316.1 | XP_011532618.1 | 16 | 2745 |