Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286734.1 | NP_001273663.1 | 3 | 303 | ||
NM_001813.2 | NP_001804.2 | 3 | 303 | |||
XM_011531544.2 | XP_011529846.1 | 3 | 296 | |||
XM_011531545.2 | XP_011529847.1 | 3 | 302 | |||
XM_011531546.2 | XP_011529848.1 | 3 | 302 | |||
XM_011531547.2 | XP_011529849.1 | 3 | 302 | |||
XM_011531548.2 | XP_011529850.1 | 3 | 302 | |||
XM_011531549.2 | XP_011529851.1 | 3 | 302 | |||
XM_017007659.1 | XP_016863148.1 | 3 | 302 | |||
GenBank mRNA | AB209996.1 | - | 3 | 228 | ||
AK290362.1 | - | 3 | 304 | |||
AU100312.1 | - | 3 | 244 | |||
Z15005.1 | - | 3 | 303 |