Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193503.1 | NP_001180432.1 | 6 | 717 | ||
NM_001976.4 | NP_001967.3 | 8 | 888 | |||
NM_053013.3 | NP_443739.3 | 8 | 846 | |||
XM_005256521.2 | XP_005256578.1 | 8 | 1274 | |||
XM_011523729.1 | XP_011522031.1 | 8 | 864 | |||
XM_017024346.1 | XP_016879835.1 | 8 | 1281 | |||
GenBank mRNA | AK300662.1 | - | 8 | 732 | ||
AK300673.1 | - | 7 | 708 | |||
AK300709.1 | - | 6 | 687 | |||
AK309864.1 | - | 9 | 939 | |||
BC017249.2 | - | 8 | 803 | 4902149 | ||
X16504.1 | - | 7 | 750 | |||
X51957.1 | - | 7 | 762 |