Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204171.1 | NP_001191100.1 | 10 | 1141 | ||
NM_001204172.1 | NP_001191101.1 | 3 | 313 | |||
NM_001278516.1 | NP_001265445.1 | 10 | 1223 | |||
NM_001278517.1 | NP_001265446.1 | 8 | 997 | |||
NM_001278518.1 | NP_001265447.1 | 7 | 812 | |||
NM_001278519.1 | NP_001265448.1 | 5 | 622 | |||
NM_002393.4 | NP_002384.2 | 11 | 1291 | |||
XM_017001311.1 | XP_016856800.1 | 11 | 1290 | |||
XM_017001312.1 | XP_016856801.1 | 11 | 1275 | |||
XM_017001313.1 | XP_016856802.1 | 10 | 1140 | |||
GenBank mRNA | AB209383.1 | - | 2 | 2855 | ||
AF007111.1 | - | 11 | 1241 | |||
AK223228.1 | - | 11 | 1254 | |||
AK304698.1 | - | 9 | 1047 | |||
AK310607.1 | - | 11 | 1267 | |||
AY923176.1 | - | 3 | 201 | |||
AY949028.1 | - | 9 | 1057 | |||
BC067299.1 | - | 11 | 1281 | 30390159 | ||
BC105106.1 | - | 10 | 1034 | 8144109 | ||
BC143431.1 | - | 10 | 1116 | |||
BC143432.1 | - | 11 | 1213 | |||
BP334671.1 | - | 11 | ||||
BX640923.1 | - | 11 | 1236 |