Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318193.1 | NP_001305122.1 | 2, 3 | 410 | ||
NM_001318194.1 | NP_001305123.1 | 3, 4 | 429 | |||
NM_002528.6 | NP_002519.1 | 3, 4 | 581 | |||
XM_017023253.1 | XP_016878742.1 | 3, 4 | 584 | |||
GenBank mRNA | AB001575.1 | - | 3, 4 | 545 | ||
BC000391.2 | - | 3, 4 | 538 | |||
BC003014.1 | - | 3, 4 | 538 | 2821314 | ||
BE900738.1 | - | 2, 3 | 292 | |||
BG468617.1 | - | 3, 4 | 355 | |||
U79718.1 | - | 3, 4 | 578 | |||
U81285.1 | - | 3, 4 | 543 | |||
Y09687.1 | - | 3, 4 | 532 |