Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164269.1 | NP_001157741.1 | 2, 3 | 239 | ||
NM_001164270.1 | NP_001157742.1 | 2, 3 | 394 | |||
NM_002875.4 | NP_002866.2 | 2, 3 | 394 | |||
NM_133487.3 | NP_597994.3 | 2, 3 | 394 | |||
XM_006720626.3 | XP_006720689.1 | 2, 3 | 445 | |||
XM_011521857.2 | XP_011520159.1 | 2, 3 | 144 | |||
XM_011521858.2 | XP_011520160.1 | 2, 3 | 191 | |||
XM_011521859.2 | XP_011520161.1 | 2, 3 | 328 | |||
XM_011521860.2 | XP_011520162.1 | 2, 3 | 239 | |||
XM_011521861.2 | XP_011520163.1 | 2, 3 | 144 | |||
XM_011521862.2 | XP_011520164.1 | 2, 3 | 362 | |||
GenBank mRNA | AK131299.1 | - | 2, 3 | 239 | ||
AK291969.1 | - | 2, 3 | 349 | |||
AK303089.1 | - | 2, 3 | 350 | |||
AK313503.1 | - | 2, 3 | 349 | |||
BC001459.2 | - | 2, 3 | 351 | 3139011 | ||
BT019705.1 | - | 1, 2 | 95 | |||
CR536559.1 | - | 1, 2 | 95 | |||
D13804.1 | - | 2, 3 | 306 | |||
D14134.1 | - | 2, 3 | 327 | |||
EU362635.1 | - | 2, 3 | 394 |