Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002812.4 | NP_002803.2 | 4 | 714 | ||
GenBank mRNA | AK301771.1 | - | 4 | 725 | ||
AK304486.1 | - | 3, 4 | 299 | |||
AK308809.1 | - | 4 | 803 | |||
BC001164.1 | - | 4 | 465 | 3528096 | ||
BC065006.1 | - | 4 | 453 | 6055235 | ||
BI560993.1 | - | 4 | 699 | |||
BQ684248.1 | - | 4 | 477 | |||
BT019688.1 | - | 4 | 383 | |||
D38047.1 | - | 4 | 453 | |||
EU668354.1 | - | 4 | 714 |