Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TJP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301025.1 | NP_001287954.1 | 6 | 614 | ||
NM_001301026.1 | NP_001287955.1 | 6 | 629 | |||
NM_003257.4 | NP_003248.3 | 5 | 850 | |||
NM_175610.3 | NP_783297.2 | 5 | 850 | |||
XM_005254617.3 | XP_005254674.2 | 6 | 734 | |||
XM_005254618.3 | XP_005254675.1 | 5 | 481 | |||
XM_005254619.3 | XP_005254676.2 | 6 | 734 | |||
XM_005254620.3 | XP_005254677.2 | 6 | 734 | |||
XM_011521972.2 | XP_011520274.2 | 6 | 734 | |||
XM_017022521.1 | XP_016878010.1 | 6 | 734 | |||
XM_017022522.1 | XP_016878011.1 | 6 | 734 | |||
XM_017022523.1 | XP_016878012.1 | 6 | 734 | |||
XM_017022524.1 | XP_016878013.1 | 6 | 734 | |||
XM_017022525.1 | XP_016878014.1 | 5 | 496 | |||
XM_017022526.1 | XP_016878015.1 | 5 | 481 | |||
XM_017022527.1 | XP_016878016.1 | 5 | 522 | |||
XM_017022528.1 | XP_016878017.1 | 5 | 496 | |||
XM_017022529.1 | XP_016878018.1 | 5 | 481 | |||
XM_017022530.1 | XP_016878019.1 | 5 | 481 | |||
XM_017022531.1 | XP_016878020.1 | 6 | 510 | |||
GenBank mRNA | AB370197.1 | - | 4 | 354 | ||
AK304758.1 | - | 5 | 481 | |||
BC111712.1 | - | 5 | 752 | 40037646 | ||
BX640879.1 | - | 6 | 630 | |||
CN372462.1 | - | 6 | 616 |