Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281297.1 | NP_001268226.1 | 3, 4 | 368 | ||
NM_003792.3 | NP_003783.1 | 3, 4 | 368 | |||
NM_153200.2 | NP_694880.1 | 3, 4 | 368 | |||
GenBank mRNA | AB002282.1 | - | 3, 4 | 327 | ||
AB002283.2 | - | 3, 4 | 327 | |||
BC015500.1 | - | 3, 4 | 318 | 3903539 | ||
BI833652.1 | - | 3, 4 | 287 | |||
BM800379.1 | - | 3, 4 | 330 | |||
BQ279031.1 | - | 3, 4 | 317 | |||
BT009863.1 | - | 3, 4 | 290 | |||
CR541914.1 | - | 3, 4 | 290 | |||
HY029111.1 | - | 3, 4 | 368 |