Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320715.1 | NP_001307644.1 | 3 | 1386 | ||
NM_003921.4 | NP_003912.1 | 3 | 1419 | |||
XM_011542397.2 | XP_011540699.1 | 3 | 1288 | |||
XM_011542398.2 | XP_011540700.1 | 3 | 1260 | |||
XM_011542399.2 | XP_011540701.1 | 3 | 1441 | |||
GenBank mRNA | AF057700.1 | - | 3 | 1114 | ||
AF082283.1 | - | 4 | 1573 | |||
AF100338.1 | - | 3 | 1114 | |||
AF134395.1 | - | 3 | 1112 | |||
AJ006288.1 | - | 4 | 1557 | |||
AK291346.1 | - | 3 | 1077 | |||
BC053617.1 | - | 3 | 989 | 6143423 | ||
BX647306.1 | - | 4 | 967 |