Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257191.2 | NP_001244120.1 | 2, 3 | 560 | ||
NM_004397.5 | NP_004388.2 | 2, 3 | 632 | |||
XM_005271417.3 | XP_005271474.1 | 2, 3 | 577 | |||
XM_011542644.1 | XP_011540946.1 | 2, 3 | 632 | |||
XM_011542645.1 | XP_011540947.1 | 2, 3 | 632 | |||
XM_017017251.1 | XP_016872740.1 | 2, 3 | 577 | |||
GenBank mRNA | AK313244.1 | - | 2, 3 | 560 | ||
BC039826.1 | - | 2, 3 | 516 | |||
BC065007.1 | - | 1, 2 | 491 | 6163439 | ||
D17532.1 | - | 2, 3 | 505 | |||
Z11685.1 | - | 2, 3 | 532 |