Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004467.3 | NP_004458.3 | 3, 4 | 350 | ||
NM_147203.2 | NP_671736.2 | 3, 4 | 438 | |||
NM_201552.1 | NP_963846.1 | 4, 5 | 477 | |||
NM_201553.1 | NP_963847.1 | 4, 5 | 565 | |||
XM_017013232.1 | XP_016868721.1 | 4, 5 | 1327 | |||
XM_017013233.1 | XP_016868722.1 | 2, 3 | 402 | |||
GenBank mRNA | AF042354.1 | - | 3, 4 | 385 | ||
AF168954.1 | - | 3, 4 | 373 | |||
AK225616.1 | - | 3, 4 | 336 | |||
AK225619.1 | - | 3, 4 | 325 | |||
AK289448.1 | - | 3, 4 | 336 | |||
AL531132.3 | - | 3, 4 | 716 | |||
AX772838.1 | - | 4, 5 | 430 | |||
BC007047.1 | - | 3, 4 | 346 | 4071810 | ||
BG565379.1 | - | 4, 5 | 503 | |||
BG566131.1 | - | 4, 5 | 460 | |||
BG567956.1 | - | 3, 4 | 441 | |||
BG618053.1 | - | 3, 4 | 351 | |||
BT006635.1 | - | 2, 3 | 241 | |||
D14446.1 | - | 3, 4 | 319 | |||
D87342.1 | - | 2, 3 | 246 |