Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004467.3 | NP_004458.3 | 5, 6 | 611 | ||
NM_147203.2 | NP_671736.2 | 5, 6 | 699 | |||
NM_201552.1 | NP_963846.1 | 6, 7 | 738 | |||
NM_201553.1 | NP_963847.1 | 6, 7 | 826 | |||
XM_017013232.1 | XP_016868721.1 | 6, 7 | 1588 | |||
XM_017013233.1 | XP_016868722.1 | 4, 5 | 663 | |||
GenBank mRNA | AF042354.1 | - | 5, 6 | 647 | ||
AF168954.1 | - | 5, 6 | 634 | |||
AK225616.1 | - | 5, 6 | 597 | |||
AK225619.1 | - | 5, 6 | 586 | |||
AK289448.1 | - | 5, 6 | 597 | |||
AX772838.1 | - | 6, 7 | 691 | |||
BC007047.1 | - | 5, 6 | 607 | 4071810 | ||
BG618053.1 | - | 5, 6 | 612 | |||
BT006635.1 | - | 4, 5 | 502 | |||
D14446.1 | - | 5, 6 | 580 | |||
D87342.1 | - | 4, 5 | 507 |