Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AXIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004655.3 | NP_004646.3 | 11 | 3184 | ||
XM_011525319.2 | XP_011523621.1 | 11 | 3395 | |||
XM_011525320.1 | XP_011523622.1 | 11 | 3384 | |||
XM_011525321.2 | XP_011523623.1 | 11 | 3332 | |||
XM_017025192.1 | XP_016880681.1 | 11 | 3051 | |||
XM_017025193.1 | XP_016880682.1 | 10 | 3189 | |||
XM_017025194.1 | XP_016880683.1 | 10 | 2968 | |||
GenBank mRNA | AB052751.1 | - | 4 | 809 | ||
AF078165.1 | - | 10 | 2983 | |||
CR933657.1 | - | 11 | 3209 | |||
CX756263.1 | - | 1 | 183 | |||
CX759580.1 | - | 2 | 564 |