Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256910.1 | NP_001243839.1 | 5, 6 | 1128 | ||
NM_004728.3 | NP_004719.2 | 5, 6 | 1009 | |||
XM_011540336.1 | XP_011538638.1 | 5, 6 | 1001 | |||
XM_017016910.1 | XP_016872399.1 | 5, 6 | 919 | |||
GenBank mRNA | AK025629.1 | - | 2, 3 | 311 | ||
AK315585.1 | - | 5, 6 | 958 | |||
BC008071.2 | - | 5, 6 | 947 | 2987980 | ||
BX648405.1 | - | 5, 6 | 895 | |||
CR749598.1 | - | 5, 6 | 1129 | |||
U41387.1 | - | 5, 6 | 960 |