Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256560.1 | NP_001243489.1 | 7 | 940 | ||
NM_004944.3 | NP_004935.1 | 8 | 1030 | |||
GenBank mRNA | AB209613.1 | - | 4 | 4094 | ||
AF047354.1 | - | 8 | 1003 | |||
AK131323.1 | - | 10 | 2062 | |||
AK301263.1 | - | 7 | 906 | |||
BC015831.1 | - | 8 | 1000 | 4723725 | ||
DB164811.1 | - | 5 | 527 | |||
U56814.1 | - | 8 | 957 |