Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005435.3 | NP_005426.2 | 6 | 3771 | ||
GenBank mRNA | AK090570.1 | - | 6 | 1383 | ||
AK123596.1 | - | 5 | 1970 | |||
AK160365.1 | - | 6 | 3827 | |||
BC010046.2 | - | 6 | 716 | |||
BC011839.2 | - | 6 | 716 | |||
BC014555.2 | - | 6 | 716 | |||
BC136661.1 | - | 6 | 3685 | 9020651 | ||
BX649178.1 | - | 5 | 1656 | |||
CR749203.1 | - | 6 | 3711 | |||
U02082.1 | - | 4 | 512 |
Gene Symbol: ARHGEF34P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033942.1 | 5 | 2689 |