Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164267.1 | NP_001157739.1 | 6 | 795 | ||
NM_005452.5 | NP_005443.3 | 6 | 957 | |||
XM_017011484.1 | XP_016866973.1 | 6 | 671 | |||
XM_017011485.1 | XP_016866974.1 | 4 | 319 | |||
XM_017011486.1 | XP_016866975.1 | 3 | 174 | |||
XM_017011487.1 | XP_016866976.1 | 4 | 319 | |||
GenBank mRNA | AF447870.1 | - | 6 | 649 | ||
AK292171.1 | - | 6 | 940 | |||
AK292616.1 | - | 6 | 740 | |||
AK297925.1 | - | 6 | 552 | |||
AK298151.1 | - | 6 | 506 | |||
AK308534.1 | - | 5 | 433 | |||
AK311388.1 | - | 5 | 654 | |||
AK315616.1 | - | 6 | 669 | |||
BC000388.2 | - | 6 | 644 | 2821198 | ||
BX439421.2 | - | 6 | 644 |