Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024959.2 | NP_001020130.1 | 3 | 351 | ||
NM_001024960.2 | NP_001020131.1 | 3 | 206 | |||
NM_001198780.1 | NP_001185709.1 | 3 | 205 | |||
NM_005718.4 | NP_005709.1 | 3 | 717 | |||
GenBank mRNA | AF006087.1 | - | 3 | 158 | ||
AF019888.1 | - | 2 | 143 | |||
AW408422.1 | - | 3 | 164 | |||
BC012596.2 | - | 2 | 140 | |||
BG938423.1 | - | 3 | 189 | |||
BI768875.1 | - | 3 | 330 | |||
BI910730.1 | - | 3 | 725 | |||
BM045872.1 | - | 3 | 216 | |||
BQ688329.1 | - | 3 | 289 | |||
BX419672.2 | - | 3 | 205 | |||
DA226584.1 | - | 3 | 206 | |||
DB060690.1 | - | 3 | 351 |
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 3 | 717 | ||
GenBank mRNA | AK024110.1 | - | 3 | 707 | ||
BC025289.1 | - | 2 | 151 |