Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005883.2 | NP_005874.1 | 3 | 372 | ||
XM_005259475.2 | XP_005259532.1 | 3 | 256 | |||
XM_006722607.2 | XP_006722670.1 | 3 | 256 | |||
XM_006722608.3 | XP_006722671.2 | 3 | 517 | |||
XM_006722609.3 | XP_006722672.1 | 3 | 326 | |||
XM_006722610.3 | XP_006722673.2 | 3 | 516 | |||
GenBank mRNA | AB012162.1 | - | 2 | 1877 | ||
AF110334.1 | - | 3 | 227 | |||
AF128222.1 | - | 2 | 368 | |||
AJ012652.1 | - | 2 | 368 | |||
AL078616.1 | - | 3 | 233 | |||
BC020381.1 | - | 3 | 226 | |||
BC032573.1 | - | 3 | 372 |