Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005883.2 | NP_005874.1 | 15 | 10033 | ||
XM_005259475.2 | XP_005259532.1 | 15 | 9917 | |||
XM_006722607.2 | XP_006722670.1 | 15 | 9914 | |||
XM_006722608.3 | XP_006722671.2 | 15 | 10178 | |||
XM_006722609.3 | XP_006722672.1 | 15 | 9987 | |||
XM_006722610.3 | XP_006722673.2 | 15 | 10174 | |||
GenBank mRNA | AB012162.1 | - | 14 | 11537 |