Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006308.2 | NP_006299.1 | 1 | 489 | ||
GenBank mRNA | AK314215.1 | - | 1 | 490 | ||
BC126272.1 | - | 1 | 344 | 8991988 | ||
BC132869.1 | - | 1 | 356 | 40146891 | ||
BX106905.1 | - | 1 | 474 | |||
CR450314.1 | - | 1 | 300 | |||
U15590.1 | - | 1 | 475 | |||
Y17782.1 | - | 1 | 448 |