Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006308.2 | NP_006299.1 | 1 | 629 | ||
GenBank mRNA | AK314215.1 | - | 1 | 630 | ||
BC126272.1 | - | 1 | 484 | 8991988 | ||
BC132869.1 | - | 1 | 496 | 40146891 | ||
CR450314.1 | - | 1 | 440 | |||
U15590.1 | - | 1 | 615 | |||
Y17782.1 | - | 1 | 588 |