Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009932.2 | NP_001009932.1 | 10 | 1759 | ||
NM_001009933.2 | NP_001009933.1 | 9 | 1501 | |||
NM_001009934.2 | NP_001009934.1 | 8 | 1416 | |||
NM_001303620.1 | NP_001290549.1 | 8 | 1382 | |||
NM_006730.3 | NP_006721.1 | 8 | 1936 | |||
XM_005277829.4 | XP_005277886.1 | 10 | 1893 | |||
XM_011531122.2 | XP_011529424.1 | 9 | 2442 | |||
XM_017029332.1 | XP_016884821.1 | 10 | 2965 | |||
GenBank mRNA | BC001561.1 | - | 8 | 1300 | 3456429 | |
BC028092.1 | - | 8 | 1936 | 5219882 | ||
L40823.1 | - | 9 | 1461 | |||
U06846.1 | - | 8 | 1318 | |||
X90392.1 | - | 9 | 1922 |