Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006773.3 | NP_006764.3 | 4 | 683 | ||
GenBank mRNA | AK001467.1 | - | 4 | 655 | ||
AK091227.1 | - | 2 | 148 | |||
BC001238.1 | - | 4 | 612 | 3452935 | ||
BC003360.1 | - | 4 | 642 | 2900252 | ||
BC024739.1 | - | 4 | 657 | 4995871 | ||
CR457060.1 | - | 3 | 404 | |||
X98743.1 | - | 3 | 421 |