Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317960.1 | NP_001304889.1 | 10 | 1178 | ||
NM_012255.4 | NP_036387.2 | 10 | 1018 | |||
XM_017027722.1 | XP_016883211.1 | 10 | 1173 | |||
XM_017027723.1 | XP_016883212.1 | 10 | 947 | |||
XM_017027724.1 | XP_016883213.1 | 10 | 1173 | |||
GenBank mRNA | AF064257.1 | - | 10 | 971 | ||
AF152169.1 | - | 10 | 921 | |||
AK001573.1 | - | 10 | 988 | |||
AK302846.1 | - | 10 | 882 | |||
AK303312.1 | - | 9 | 829 | |||
AK314602.1 | - | 10 | 957 | |||
AL136841.1 | - | 10 | 944 | |||
AY382900.1 | - | 9 | 833 |