Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013325.4 | NP_037457.3 | 13 | 1924 | ||
NM_178326.2 | NP_847896.1 | 13 | 1944 | |||
XM_005246992.3 | XP_005247049.2 | 12 | 2695 | |||
XM_005246993.2 | XP_005247050.2 | 13 | 1946 | |||
GenBank mRNA | AB023160.2 | - | 13 | 1831 | ||
AK027763.1 | - | 13 | 1877 | |||
AK123425.1 | - | 4 | 1291 | |||
AK125277.1 | - | 13 | 2368 | |||
AL050288.1 | - | 1 | 724 | |||
AL133638.1 | - | 1 | 25 | |||
BC000719.2 | - | 13 | 1829 | 3504267 |