Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293186.1 | NP_001280115.1 | 5 | 655 | ||
NM_001293187.1 | NP_001280116.1 | 5 | 569 | |||
NM_014168.3 | NP_054887.2 | 4 | 796 | |||
XM_011511023.1 | XP_011509325.1 | 5 | 596 | |||
GenBank mRNA | AF161482.1 | - | 5 | 541 | ||
AF201938.1 | - | 5 | 613 | |||
AK001321.1 | - | 5 | 543 | |||
AL523293.3 | - | 4 | 778 | |||
BC000921.2 | - | 4 | 708 | 3445582 | ||
BC093014.1 | - | 5 | 551 | 4799279 |