Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014448.3 | NP_055263.2 | 11 | 1797 | ||
XM_017001049.1 | XP_016856538.1 | 11 | 2398 | |||
XM_017001050.1 | XP_016856539.1 | 11 | 1603 | |||
XM_017001051.1 | XP_016856540.1 | 11 | 1808 | |||
GenBank mRNA | AK091910.1 | - | 5 | 836 | ||
AK095984.1 | - | 11 | 1704 | |||
AK296150.1 | - | 6 | 695 | |||
BC002681.1 | - | 10 | 1084 | 3613623 | ||
BC051838.1 | - | 9 | 1104 | 6191377 | ||
BT007270.1 | - | 6 | 685 | |||
D89016.1 | - | 8 | 1112 |