Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014448.3 | NP_055263.2 | 12 | 1890 | ||
XM_017001049.1 | XP_016856538.1 | 12 | 2491 | |||
XM_017001050.1 | XP_016856539.1 | 12 | 1696 | |||
XM_017001051.1 | XP_016856540.1 | 12 | 1901 | |||
GenBank mRNA | AK091910.1 | - | 5 | 1326 | ||
AK095984.1 | - | 12 | 1797 | |||
AK296150.1 | - | 7 | 788 | |||
BC002681.1 | - | 11 | 1177 | 3613623 | ||
BC051838.1 | - | 10 | 1197 | 6191377 | ||
BT007270.1 | - | 7 | 778 | |||
D89016.1 | - | 9 | 1205 |