Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014448.3 | NP_055263.2 | 15 | 2722 | ||
XM_017001049.1 | XP_016856538.1 | 15 | 3321 | |||
XM_017001050.1 | XP_016856539.1 | 15 | 2526 | |||
XM_017001051.1 | XP_016856540.1 | 15 | 2731 | |||
GenBank mRNA | AK091910.1 | - | 8 | 2158 | ||
AK095984.1 | - | 15 | 2629 | |||
BC002681.1 | - | 14 | 2009 | 3613623 | ||
BC051838.1 | - | 13 | 2029 | 6191377 | ||
D89016.1 | - | 12 | 2037 |